Merck
  • Elimination reactions of (E)-2, 4-dinitrobenzaldehyde O-aryloximes promoted by RO-/ROH buffers in EtOH.
Elimination reactions of (E)-2, 4-dinitrobenzaldehyde O-aryloximes promoted by RO-/ROH buffers in EtOH.
The Journal of Organic Chemistry (1998)
Cho BR, et al.