- Deficiency of protein C in congenital thrombotic disease.
Deficiency of protein C in congenital thrombotic disease.
The Journal of clinical investigation (1981-11-01)
J H Griffin, B Evatt, T S Zimmerman, A J Kleiss, C Wideman
PMID6895379
ZUSAMMENFASSUNG
A family with a history of recurring thrombosis was studied to determine if a plasma protein deficiency could account for the observed disease. Protein C levels in plasma were determined immunologically using the Laurell rocket technique. The propositus, his father, and his paternal uncle, who are severely affected, had 38-49% of normal levels of protein C antigen, whereas unaffected family members had normal levels. There was no familial deficiency of antithrombin III and plasminogen. Because activated protein C is a potent in vitro anticoagulant enzyme and an in vivo profibrinolytic agent, it is suggested that the recurrent thrombotic disease in this family is due to an inherited deficiency in protein C.