Merck
  • LuxR homologue SmcR is essential for Vibrio vulnificus pathogenesis and biofilm detachment, and its expression is induced by host cells.
LuxR homologue SmcR is essential for Vibrio vulnificus pathogenesis and biofilm detachment, and its expression is induced by host cells.
Infection and immunity (2013-07-31)
Seung Min Kim, Jin Hwan Park, Hyun Sung Lee, Won Bin Kim, Jung Min Ryu, Ho Jae Han, Sang Ho Choi
ZUSAMMENFASSUNG
Quorum sensing is a cell-to-cell communication system known to control many bacterial processes. In the present study, the functions of quorum sensing in the pathogenesis of Vibrio vulnificus, a food-borne pathogen, were assessed by evaluating the virulence of a mutant deficient in SmcR, a quorum-sensing regulator and homologue of LuxR. When biofilms were used as an inoculum, the smcR mutant was impaired in virulence and colonization capacity in the infection of mice. The lack of SmcR also resulted in decreased histopathological damage in mouse jejunum tissue. These results indicated that SmcR is essential for V. vulnificus pathogenesis. Moreover, the smcR mutant exhibited significantly reduced biofilm detachment. Upon exposure to INT-407 host cells, the wild type, but not the smcR mutant, revealed accelerated biofilm detachment. The INT-407 cells increased smcR expression by activating the expression of LuxS, an autoinducer-2 synthase, indicating that host cells manipulate the cellular level of SmcR through the quorum-sensing signaling of V. vulnificus. A whole-genome microarray analysis revealed that the genes primarily involved in biofilm detachment and formation are up- and downregulated by SmcR, respectively. Among the SmcR-regulated genes, vvpE encoding an elastolytic protease was the most upregulated, and the purified VvpE appeared to dissolve established biofilms directly in a concentration-dependent manner in vitro. These results suggest that the host cell-induced SmcR enhances the detachment of V. vulnificus biofilms entering the host intestine and thereby may promote the dispersal of the pathogen to new colonization loci, which is crucial for pathogenesis.