Merck
  • Restriction endonuclease Aor13HI from Acidiphilium organovorum 13H, a new isoschizomer of BspMII: purification and characterization.
Restriction endonuclease Aor13HI from Acidiphilium organovorum 13H, a new isoschizomer of BspMII: purification and characterization.
Bioscience, biotechnology, and biochemistry (1993-10-01)
K Inagaki, T Hikita, S Yanagidani, Y Nomura, N Kishimoto, T Tano, H Tanaka
ZUSAMMENFASSUNG
A restriction endonuclease, Aor13HI, an isoschizomer of BspMII, was purified to homogeneity from cell extracts of Acidiphilium organovorum strain 13H. The enzyme has a molecular mass of 60,000 daltons and consists of two subunits identical in molecular mass of 30,000 daltons. Aor13HI endonuclease, like BspMII, recognizes the palindromic six-base sequence 5'-TCCGGA-3', and cleaves between the T and C to produce a four-base 5' extension. Aor13HI is not inhibited by dam-dependent methylation. The isoelectric point of the enzyme is 5.7. Aor13HI activity was maximum at pH 7.5, 100 mM KCl, 7.5-10 mM MgCl2, and 55 degrees C. The enzyme was stable up to 60 degrees C. The N-terminal amino acid sequence (30 residues) of Aor13HI did not show any similarity with the sequence of other restriction endonucleases reported.