Merck
The BCL11A transcription factor directly activates RAG gene expression and V(D)J recombination.
Molecular and cellular biology (2013-02-27)
Baeck-seung Lee, Joseph D Dekker, Bum-kyu Lee, Vishwanath R Iyer, Barry P Sleckman, Arthur L Shaffer, Gregory C Ippolito, Philip W Tucker
ZUSAMMENFASSUNG
Recombination-activating gene 1 protein (RAG1) and RAG2 are critical enzymes for initiating variable-diversity-joining (VDJ) segment recombination, an essential process for antigen receptor expression and lymphocyte development. The transcription factor BCL11A is required for B cell development, but its molecular function(s) in B cell fate specification and commitment is unknown. We show here that the major B cell isoform, BCL11A-XL, binds the RAG1 promoter and Erag enhancer to activate RAG1 and RAG2 transcription in pre-B cells. We employed BCL11A overexpression with recombination substrates in a cultured pre-B cell line as well as Cre recombinase-mediated Bcl11a(lox/lox) deletion in explanted murine pre-B cells to demonstrate direct consequences of BCL11A/RAG modulation on V(D)J recombination. We conclude that BCL11A is a critical component of a transcriptional network that regulates B cell fate by controlling V(D)J recombination.