Merck
  • PDGFRB-rearranged T-lymphoblastic leukemia/lymphoma occurring with myeloid neoplasms: the missing link supporting a stem cell origin.
PDGFRB-rearranged T-lymphoblastic leukemia/lymphoma occurring with myeloid neoplasms: the missing link supporting a stem cell origin.
Haematologica (2014-06-22)
Sarah L Ondrejka, Armin G Jegalian, Annette S Kim, Devon S Chabot-Richards, Jennifer Giltnane, David R Czuchlewski, Shashirekha Shetty, Mikkael A Sekeres, Ashwini Yenamandra, David Head, Madan Jagasia, Eric D Hsi