Merck
  • The Activators of Cyclin-Dependent Kinase 5 p35 and p39 Are Essential for Oligodendrocyte Maturation, Process Formation, and Myelination.
The Activators of Cyclin-Dependent Kinase 5 p35 and p39 Are Essential for Oligodendrocyte Maturation, Process Formation, and Myelination.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2016-03-11)
Fucheng Luo, Jessie Zhang, Kathryn Burke, Robert H Miller, Yan Yang
ZUSAMMENFASSUNG
The regulation of oligodendrocyte development and myelin formation in the CNS is poorly defined. Multiple signals influence the rate and extent of CNS myelination, including the noncanonical cyclin-dependent kinase 5 (Cdk5) whose functions are regulated by its activators p35 and p39. Here we show that selective loss of either p35 or p39 perturbed specific aspects of oligodendrocyte development, whereas loss of both p35 and p39 completely inhibited the development of mature oligodendrocytes and myelination. In the absence of p35, oligodendrocyte differentiation was delayed, process outgrowth was truncated in vitro, and the patterning and extent of myelination were perturbed in the CNS of p35(-/-) mice. In the absence of p39, oligodendrocyte maturation was transiently affected both in vitro and in vivo. However, loss of both p35 and p39 in oligodendrocyte lineage cells completely inhibited oligodendrocyte progenitor cell differentiation and myelination both in vitro and after transplantation into shiverer slice cultures. Loss of p35 and p39 had a more profound effect on oligodendrocyte development than simply the loss of Cdk5 and could not be rescued by Cdk5 overexpression. These data suggest p35 and p39 have specific and overlapping roles in oligodendrocyte development, some of which may be independent of Cdk5 activation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluidinblau, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
Bernsteinsäure bis(N-Hydroxysuccinimid-Ester), ≥95%, powder
Sigma-Aldrich
Histon H1-Protein, 20 mg, Purified bovine Histone H1 protein for use as a substrate in histone modification assays (HAT, HDAC, DNMT) and chromatin assembly studies.