Merck
  • Bone marrow stroma-induced resistance of chronic lymphocytic leukemia cells to arsenic trioxide involves Mcl-1 upregulation and is overcome by inhibiting the PI3Kδ or PKCβ signaling pathways.
Bone marrow stroma-induced resistance of chronic lymphocytic leukemia cells to arsenic trioxide involves Mcl-1 upregulation and is overcome by inhibiting the PI3Kδ or PKCβ signaling pathways.
Oncotarget (2015-11-06)
Irene Amigo-Jiménez, Elvira Bailón, Noemí Aguilera-Montilla, María José Terol, José A García-Marco, Angeles García-Pardo
ZUSAMMENFASSUNG
CLL remains an incurable disease in spite of the many new compounds being studied. Arsenic trioxide (ATO) induces apoptosis in all CLL cell types and could constitute an efficient therapy. To further explore this, we have studied the influence of stromal cells, key components of the CLL microenvironment, on the response of CLL cells to ATO. Bone marrow stromal cells induced CLL cell resistance to 2 μM ATO and led to activation of Lyn, ERK, PI3K and PKC, as well as NF-κB and STAT3. Mcl-1, Bcl-xL, and Bfl-1 were also upregulated after the co-culture. Inhibition experiments indicated that PI3K and PKC were involved in the resistance to ATO induced by stroma. Moreover, idelalisib and sotrastaurin, specific inhibitors for PI3Kδ and PKCβ, respectively, inhibited Akt phosphorylation, NF-κB/STAT3 activation and Mcl-1 upregulation, and rendered cells sensitive to ATO. Mcl-1 was central to the mechanism of resistance to ATO, since: 1) Mcl-1 levels correlated with the CLL cell response to ATO, and 2) blocking Mcl-1 expression or function with specific siRNAs or inhibitors overcame the protecting effect of stroma. We have therefore identified the mechanism involved in the CLL cell resistance to ATO induced by bone marrow stroma and show that idelalisib or sotrastaurin block this mechanism and restore sensibility to ATO. Combination of ATO with these inhibitors may thus constitute an efficient treatment for CLL.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Vinculin-Antikörper in Maus hergestellte Antikörper, clone hVIN-1, ascites fluid
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, ascites fluid
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
Triciribine hydrate, ≥97% (HPLC)
Sigma-Aldrich
NF-κB-Aktivierungsinhibitor, InSolution, ≥98%
Sigma-Aldrich
GF 109203X, synthetic, ≥90% (HPLC)