Merck
MicroRNA-203 suppresses gastric cancer growth by targeting PIBF1/Akt signaling.
Journal of experimental & clinical cancer research : CR (2016-03-17)
Shao-Jun Chu, Ge Wang, Peng-Fei Zhang, Rui Zhang, Yan-Xia Huang, Yun-Min Lu, Wei Da, Qun Sun, Jing Zhang, Jin-Shui Zhu
ZUSAMMENFASSUNG
MicroRNAs (miRNAs) have been proved involved in many tumorigenic behaviors including tumor growth. But, the clinical significance and functions of miRNA-203 in gastric cancer (GC) remain elusive. Decreased expression of miRNA-203 was correlated with tumor size, poor prognosis and recurrence in GC patients. Overexpression of miR-203 or knockdown of its target progesterone immunomodulatory binding factor 1 (PIBF1) inhibited GC growth in vitro and in vivo, while miR-203 knockdown promoted GC proliferation. In addition, PIBF1 overexpression attenuated the inhibitory effects of miR-203 on GC growth and enhanced that effect on p-Akt expression. MiR-203 as a tumor biomarker suppresses GC growth through targeting the PIBF1/Akt signaling, suggesting that it may have the important therapeutic potential for the treatment of GC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human PIBF1