Merck
Effects of technological processes on enniatin levels in pasta.
Journal of the science of food and agriculture (2015-06-03)
Ana B Serrano, Guillermina Font, Jordi Mañes, Emilia Ferrer
ZUSAMMENFASSUNG
Potential human health risks posed by enniatins (ENs) require their control primarily from cereal products, creating a demand for harvesting, food processing and storage techniques capable to prevent, reduce and/or eliminate the contamination. In this study, different methodologies to pasta processing simulating traditional and industrial processes were developed in order to know the fate of the mycotoxin ENs. The levels of ENs were studied at different steps of pasta processing. The effect of the temperature during processing was evaluated in two types of pasta (white and whole-grain pasta). Mycotoxin analysis was performed by LC-MS/MS. High reductions (up to 50% and 80%) were achieved during drying pasta at 45-55°C and 70-90°C, respectively. The treatments at low temperature (25°C) did not change EN levels. The effect of pasta composition did not cause a significant effect on the stability of ENs. The effect of the temperature allowed a marked mycotoxin reduction during pasta processing. Generally, ENA1 and ENB showed higher thermal stability than did ENA and ENB1 . The findings from the present study suggested that pasta processing at medium-high temperatures is a potential tool to remove an important fraction of ENs from the initial durum wheat semolina.