Merck
  • The spatial distribution of the photostability of thionine in zeolite L nanochannels investigated by Photobleaching Lifetime Imaging Microscopy.
The spatial distribution of the photostability of thionine in zeolite L nanochannels investigated by Photobleaching Lifetime Imaging Microscopy.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2016-02-19)
Diego Lencione, Marcelo H Gehlen, Loren N Trujillo, Renan C F Leitao, Rodrigo Q Albuquerque
ZUSAMMENFASSUNG
Dye photobleaching is a photochemical reaction that can be investigated locally using fluorescence microscopy techniques. In this study, a user-friendly computational tool to assist photobleaching experiments called Photobleaching Lifetime Imaging Microscopy (PbLIM) is presented. With this tool it is possible to recover the photobleaching kinetics spatially, where a photobleaching lifetime is generated for each pixel of the image. Our model was applied to the photobleaching process of thionine encapsulated into the one-dimensional nano-channels of Zeolite L (ZL), from where we gained insight into the molecular oxygen distribution inside the ZL channels, as well as the detailed photobleaching of the confined thionine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thionin -acetat (Salz), certified by the BSC, Dye content ≥85 %