Merck
Benzimidazole derivatives as new α-glucosidase inhibitors and in silico studies.
Bioorganic chemistry (2015-12-08)
Nik Khairunissa Nik Abdullah Zawawi, Muhammad Taha, Norizan Ahmat, Abdul Wadood, Nor Hadiani Ismail, Fazal Rahim, Syed Sikander Azam, Noraishah Abdullah
ZUSAMMENFASSUNG
Newly synthesized benzimidazole hydrazone derivatives 1-26 were evaluated for their α-glucosidase inhibitory activity. Compounds 1-26 exhibited varying degrees of yeast α-glucosidase inhibitory activity with IC50 values between 8.40 ± 0.76 and 179.71 ± 1.11 μM when compared with standard acarbose. In this assay, seven compounds that showed highest inhibitory effects than the rest of benzimidazole series were identified. All the synthesized compounds were characterized by different spectroscopic methods adequately. We further evaluated the interaction of the active compounds with enzyme with the help of docking studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzimidazol, 98%