Merck
  • Development and validation of control materials for the measurement of vitamin D 3 in selected US foods
Development and validation of control materials for the measurement of vitamin D 3 in selected US foods
J. Food Compos. Anal. (2008)
Katherine Phillips