Merck
  • Nutritional quality and amino acid composition of diets consumed by scavenging hens and cocks across seasons.
Nutritional quality and amino acid composition of diets consumed by scavenging hens and cocks across seasons.
Tropical animal health and production (2016-03-05)
Cyprial Ndumiso Ncobela, Michael Chimonyo
ZUSAMMENFASSUNG
The objective of the study was to determine the effect of season on nutritional quality and amino acid composition of diets that scavenging hens and cocks consume. Thirty hens and 30 cocks were purchased and slaughtered during each of the rainy, post rainy, cool dry and hot dry seasons. A total of 240 birds were used in the study. Fresh crop content weights were high (P < 0.05) during the cool dry season. Cereal grains, kitchen wastes, green materials, animal protein sources and inorganic materials were the main components of the crop contents. Crop contents varied with season and sex of bird (P < 0.05). The cereal grain weights were high during cool dry and hot dry seasons. Weights of animal protein sources (insects, locusts and termites) were higher (P < 0.05) during the rainy and post rainy seasons. Hens contained more animal protein sources (P < 0.05) than cocks. Hens had a higher (P < 0.05) lysine content during the rainy season than cocks. Histidine, serine, arginine, threonine, cysteine and lysine contents varied with seasons (P < 0.05). Methionine did not vary with season and sex of the bird. Nutritional supplementation of village chickens should, therefore, vary with seasons.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Amylase, hitzestabil, solution, for use in Total Dietary Fiber Assay, TDF-100A