Merck
  • High-throughput quantitative profiling of serum N-glycome by MALDI-TOF mass spectrometry and N-glycomic fingerprint of liver fibrosis.
High-throughput quantitative profiling of serum N-glycome by MALDI-TOF mass spectrometry and N-glycomic fingerprint of liver fibrosis.
Clinical chemistry (2007-05-19)
Richard K T Kam, Terence C W Poon, Henry L Y Chan, Nathalie Wong, Alex Y Hui, Joseph J Y Sung
ZUSAMMENFASSUNG
The use of MALDI-TOF mass spectrometry (MS) in quantitative glycan profiling has not been reported. In this study, we attempted to establish a high-throughput quantitative assay for profiling serum N-glycome, and we applied the new assay to identifying serum N-glycans for diagnosis of liver fibrosis and cirrhosis. N-glycans from whole serum proteins in 2 microL serum were released by enzymatic digestion, cleaned up by hydrophilic chromatography, and subsequently quantitatively profiled with a linear MALDI-TOF MS system, which was originally designed for quantitative proteomic profiling. Serum N-glycome profiles from 46 patients with chronic hepatitis B infection and with different degrees of liver fibrosis were examined. The intra- and interassay CVs of peak intensities of the standard N-glycans were <8% and <17%, respectively. When the assay was applied to the analysis of serum N-glycome profiles, 17 peaks were found to be potential biomarkers for detection of liver fibrosis/cirrhosis. Linear regression analysis revealed that 4 peaks of 1341.5, 1829.7, 1933.3, and 2130.3 m/z (all P <0.005) had complementary value in detecting liver fibrosis and included them, but not any serological markers, in the diagnostic model. Leave-one-out cross-validation showed the diagnostic model could identify significant fibrosis (Ishak score > or = 3) and cirrhosis (Ishak score > or = 5), both at 85% accuracy. This is the first study to illustrate the quantitative aspect of MALDI-TOF MS in N-glycome profiling and the first study to reveal the potential value of the serum N-glycan profile for identifying liver fibrosis.