Merck
  • Preparation and a time-resolved fluoroimmunoassay application of new europium fluorescent nanoparticles.
Preparation and a time-resolved fluoroimmunoassay application of new europium fluorescent nanoparticles.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2004-04-02)
Xiaodan Hai, Mingqian Tan, Guilan Wang, Zhiqiang Ye, Jingli Yuan, Kazuko Matsumoto
ZUSAMMENFASSUNG
New silica-based europium fluorescent nanoparticles having surface amino groups were prepared by a covalent binding-copolymerization technique. In the nanoparticles, the fluorescent Eu3+ chelate molecules were covalently bound to silicon atoms to protect the nanoparticles from dye leaking in bio-applications. The amino groups on the surface of nanoparticles made the surface modification and bioconjugation of nanoparticles easier. The nanoparticles were characterized and developed as a new type of fluorescence probe for a highly sensitive time-resolved fluoroimmunoassay (TR-FIA) of human hepatitis B surface antigen (HBsAg).