Merck
Coordination polymers of 5-substituted isophthalic acid.
CrystEngComm (2016-03-29)
Laura J McCormick, Samuel A Morris, Alexandra M Z Slawin, Simon J Teat, Russell E Morris
ZUSAMMENFASSUNG
The synthesis and characterisation of five coordination polymers - Ni