Merck
  • Synthesis of isotopically labelled [3-14C]-and [3, 3-2H2]-5-fluoro-1, 3-dihydro-1-hydroxy-2, 1-benzoxaborole (AN2690), a new antifungal agent for the potential treatment of onychomycosis.
Synthesis of isotopically labelled [3-14C]-and [3, 3-2H2]-5-fluoro-1, 3-dihydro-1-hydroxy-2, 1-benzoxaborole (AN2690), a new antifungal agent for the potential treatment of onychomycosis.
Journal of Labelled Compounds & Radiopharmaceuticals (2007)
Baker SJ, et al.