- Nitrogen heterocycles as building blocks for new metallosupramolecular architectures.
Nitrogen heterocycles as building blocks for new metallosupramolecular architectures.
Molecules (Basel, Switzerland) (2007-11-17)
Peter J Steel
PMID18007443
ZUSAMMENFASSUNG
The syntheses of representative examples of five classes of new heterocyclic ligands are described. These include N,N'-chelating bis-heterocycles, binucleating ligands, cyclometallated compounds, chiral ligands and a family of polyheteroaryl-linked arenes.