Merck
  • Novel class of Bi(iii) hydroxamato complexes: synthesis, urease inhibitory activity and activity against H. pylori.
Novel class of Bi(iii) hydroxamato complexes: synthesis, urease inhibitory activity and activity against H. pylori.
Dalton transactions (Cambridge, England : 2003) (2016-06-18)
D M Keogan, B Twamley, D Fitzgerald-Hughes, D M Griffith
ZUSAMMENFASSUNG
Reaction of Bi(NO3)3 with benzohydroxamic acid (Bha) and salicylhydroxamic acid (Sha) gives the novel Bi(iii) complexes [Bi2(Bha-1H)2(μ-Bha-1H)2(η(2)-NO3)2] () and [Bi6(CH3OH)2(η(1)-NO3)2(η(2)-NO3)(OH2)2(Sha-1H)12](NO3)2 (). X-ray crystal structure of reveals two hydroxamato coordination modes; bidentate bridging (O, O') and bidentate non-bridging (O, O') and of reveals one coordination mode; bidentate bridging (O, O'). , specifically designed to and demonstrated to inhibit the activity of urease, exhibits excellent antibacterial activity against three strains of Helicobacter pylori with MIC ≥ 16 μg mL(-1).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Hydroxybenzohydroxamsäure, 99%
Sigma-Aldrich
Benzhydroxamsäure, 99%