Merck
  • Cyclic stretch promotes osteogenesis-related gene expression in osteoblast-like cells through a cofilin-associated mechanism.
Cyclic stretch promotes osteogenesis-related gene expression in osteoblast-like cells through a cofilin-associated mechanism.
Molecular medicine reports (2016-05-14)
Jie Gao, Shanmin Fu, Zhaobin Zeng, Feifei Li, Qiannan Niu, Da Jing, Xue Feng
ZUSAMMENFASSUNG
Osteoblasts have the capacity to perceive and transduce mechanical signals, and thus, regulate the mRNA and protein expression of a variety of genes associated with osteogenesis. Cytoskeletal reconstruction, as one of the earliest perception events for external mechanical stimulation, has previously been demonstrated to be essential for mechanotransduction in bone cells. However, the mechanism by which mechanical signals induce cytoskeletal deformation remains poorly understood. The actin‑binding protein, cofilin, promotes the depolymerization of actin and is understood to be important in the regulation of activities in various cell types, including endothelial, neuronal and muscle cells. However, to the best of our knowledge, the importance of cofilin in osteoblastic mechanotransduction has not been previously investigated. In the present study, osteoblast‑like MG‑63 cells were subjected to physiological cyclic stretch stimulation (12% elongation) for 1, 4, 8, 12 and 24 h, and the expression levels of cofilin and osteogenesis-associated genes were quantified with reverse transcription‑quantitative polymerase chain reaction, immunofluorescence staining and western blotting analyses. Additionally, knockdown of cofilin using RNA interference was conducted, and the mRNA levels of osteogenesis‑associated genes were compared between osteoblast‑like cells in the presence and absence of cofilin gene knockdown. The results of the present study demonstrated that cyclic stretch stimulates the expression of genes associated with osteoblastic activities in MG‑63 cells, including alkaline phosphatase (ALP), osteocalcin (OCN), runt‑related transcription factor 2 (Runx2) and collagen‑1 (COL‑1). Cyclic stretch also regulates the mRNA and protein expression of cofilin in MG‑63 cells. Furthermore, stretch‑induced increases in the levels of osteogenesis-associated genes, including ALP, OCN, Runx2 and COL‑1, were reduced following cofilin gene knockdown. Together, these results demonstrate that cofilin is involved in the regulation of mechanical load‑induced osteogenesis and, to the best of our knowledge, provides the first evidence demonstrating the importance of cofilin in osteoblastic mechanotransduction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Kaninchen-IgG-Antikörper der Ziege, (H+L) HRP-Konjugat, 1 mg/mL, Chemicon®