Merck
  • Novel indium-mediated ternary reactions between indole-3-carboxaldehydes-allyl bromide-enamines: facile synthesis of bisindolyl-and indolyl-heterocyclic alkanes.
Novel indium-mediated ternary reactions between indole-3-carboxaldehydes-allyl bromide-enamines: facile synthesis of bisindolyl-and indolyl-heterocyclic alkanes.
Tetrahedron Letters (2003)
Kumar S, et al.