- Epigenetics in cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Bacterial Glycerol Stock, Epigenetic Regulators
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Epigenetic Regulators