Merck
  • Alcohol and Aldehyde Dehydrogenases Contribute to Sex-Related Differences in Clearance of Zolpidem in Rats.
Alcohol and Aldehyde Dehydrogenases Contribute to Sex-Related Differences in Clearance of Zolpidem in Rats.
Frontiers in pharmacology (2016-08-31)
Cody J Peer, Jonathan D Strope, Shaunna Beedie, Ariel M Ley, Alesia Holly, Karim Calis, Ronald Farkas, Jagan Parepally, Angela Men, Emmanuel O Fadiran, Pamela Scott, Marjorie Jenkins, William H Theodore, Tristan M Sissung
ZUSAMMENFASSUNG
The recommended zolpidem starting dose was lowered in females (5 mg vs. 10 mg) since side effects were more frequent and severe than those of males; the mechanism underlying sex differences in pharmacokinetics (PK) is unknown. We hypothesized that such differences were caused by known sex-related variability in alcohol dehydrogenase (ADH) expression. Male, female, and castrated male rats were administered 2.6 mg/kg zolpidem, ± disulfiram (ADH/ALDH pathway inhibitor) to compare PK changes induced by sex and gonadal hormones. PK analyses were conducted in rat plasma and rat brain. Sex differences in PK were evident: females had a higher C MAX (112.4 vs. 68.1 ug/L) and AUC (537.8 vs. 231.8 h(∗)ug/L) than uncastrated males. Castration induced an earlier T MAX (0.25 vs. 1 h), greater C MAX (109.1 vs. 68.1 ug/L), and a corresponding AUC increase (339.7 vs. 231.8 h(∗)ug/L). Administration of disulfiram caused more drastic C MAX and T MAX changes in male vs. female rats that mirrored the effects of castration on first-pass metabolism, suggesting that the observed PK differences may be caused by ADH/ALDH expression. Brain concentrations paralleled plasma concentrations. These findings indicate that sex differences in zolpidem PK are influenced by variation in the expression of ADH/ALDH due to gonadal androgens.