Merck
Exploring host-pathogen interactions through genome wide protein microarray analysis.
Scientific reports (2016-06-16)
Luigi Scietti, Katia Sampieri, Irene Pinzuti, Erika Bartolini, Barbara Benucci, Alessia Liguori, Andreas F Haag, Paola Lo Surdo, Werner Pansegrau, Vincenzo Nardi-Dei, Laura Santini, Seguinde Arora, Xavier Leber, Simonetta Rindi, Silvana Savino, Paolo Costantino, Domenico Maione, Marcello Merola, Pietro Speziale, Matthew J Bottomley, Fabio Bagnoli, Vega Masignani, Mariagrazia Pizza, Meike Scharenberg, Jean-Marc Schlaeppi, Mikkel Nissum, Sabrina Liberatori
ZUSAMMENFASSUNG
During bacterial pathogenesis extensive contacts between the human and the bacterial extracellular proteomes take place. The identification of novel host-pathogen interactions by standard methods using a case-by-case approach is laborious and time consuming. To overcome this limitation, we took advantage of large libraries of human and bacterial recombinant proteins. We applied a large-scale protein microarray-based screening on two important human pathogens using two different approaches: (I) 75 human extracellular proteins were tested on 159 spotted Staphylococcus aureus recombinant proteins and (II) Neisseria meningitidis adhesin (NadA), an important vaccine component against serogroup B meningococcus, was screened against ≈2300 spotted human recombinant proteins. The approach presented here allowed the identification of the interaction between the S. aureus immune evasion protein FLIPr (formyl-peptide receptor like-1 inhibitory protein) and the human complement component C1q, key players of the offense-defense fighting; and of the interaction between meningococcal NadA and human LOX-1 (low-density oxidized lipoprotein receptor), an endothelial receptor. The novel interactions between bacterial and human extracellular proteins here presented might provide a better understanding of the molecular events underlying S. aureus and N. meningitidis pathogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-polyhistidin-Antikörper in Maus hergestellte Antikörper, clone HIS-1, ascites fluid