Merck
  • Anti-cancer activity of Psoralea fructus through the downregulation of cyclin D1 and CDK4 in human colorectal cancer cells.
Anti-cancer activity of Psoralea fructus through the downregulation of cyclin D1 and CDK4 in human colorectal cancer cells.
BMC complementary and alternative medicine (2016-09-28)
Gwang Hun Park, Ji Ho Sung, Hun Min Song, Jin Boo Jeong
ZUSAMMENFASSUNG
Psoralea Fructus (PF), the dried and ripe fruit of Psoralea corylifolia exhibits an anti-cancer activity. However, the molecular mechanisms by which PF inhibits the proliferation of cancer cells have not been elucidated in detail. Cyclin D1 and CDK4 are important regulatory proteins in cell growth and are overexpressed in many cancer cells. In this study, we investigated the molecular mechanism of PF on the downregulation of cyclin D1 and CDK4 level. Cell growth was evaluated by MTT assay. The effect of PF on cyclin D1 and CDK4 expression was evaluated by Western blot or RT-PCR. PF suppressed the proliferation of human colorectal cancer cell lines such as HCT116 (IC Our results suggest that PF has potential to be a candidate for the development of chemoprevention or therapeutic agents for human colorectal cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
SP600125, ≥98% (HPLC)