Merck
C5b-9 Staining Correlates With Clinical and Tumor Stage in Gastric Adenocarcinoma.
Applied immunohistochemistry & molecular morphology : AIMM (2015-07-18)
Jian Chen, Wei-Jun Yang, Hai-Jian Sun, Xia Yang, Yu-Zhang Wu
ZUSAMMENFASSUNG
The complement system is a critical part of the immune response, acting in defense against viral infections, clearance of immune complexes, and maintenance of tissue homeostasis. Upregulated expression of the terminal complement complex, C5b-9, has been observed on various tumor cells, such as stomach carcinoma cells, and on cells in the necrotic regions of these tumors as well; however, whether and how C5b-9 is related to gastric cancer progression and severity remains unknown. In this study, human gastric adenocarcinoma (HGAC) tissues (n=47 cases) and patient-matched adjacent nontumoral parenchyma (n=20 cases) were evaluated by tissue microarray and immunohistochemistry. The HGAC tissues showed upregulated C5b-9 expression. Multinomial logistic regression and likelihood ratio testing showed that overexpression of C5b-9 in HGAC tissue was significantly correlated with clinical stage (P=0.007) and tumor stage (P=0.005), but not with tumor distant organ metastasis, lymphoid nodal status, sex, or age. Patients with late-stage gastric adenocarcinoma had a higher amount of tumor cells showing positive staining for C5b-9 than patients with early-stage disease. These results may help in diagnosis and assessment of disease severity of human gastric carcinoma.