Merck
  • Isolation of a novel human gene, MARKL1, homologous to MARK3 and its involvement in hepatocellular carcinogenesis.
Isolation of a novel human gene, MARKL1, homologous to MARK3 and its involvement in hepatocellular carcinogenesis.
Neoplasia (New York, N.Y.) (2001-04-28)
T Kato, S Satoh, H Okabe, O Kitahara, K Ono, C Kihara, T Tanaka, T Tsunoda, Y Yamaoka, Y Nakamura, Y Furukawa
ZUSAMMENFASSUNG
Activation of the Wnt-signaling pathway is known to play a crucial role in carcinogenesis of various human organs including the colon, liver, prostate, and endometrium. To investigate the mechanisms underlying hepatocellular carcinogenesis, we attempted to identify genes regulated by beta-catenin/Tcf complex in a human hepatoma cell line, HepG2, in which an activated form of beta-catenin is expressed. By means of cDNA microarray, we isolated a novel human gene, termed MARKL1 (MAP/microtubule affinity-regulating kinase-like 1), whose expression was downregulated in response to decreased Tcf/LEF1 activity. The transcript expressed in liver consisted of 3529 nucleotides that contained an open reading frame of 2256 nucleotides, encoding 752 amino acids homologous to human MARK3 (MAP/microtubule affinity-regulating kinase 3). Expression levels of MARKL1 were markedly elevated in eight of nine HCCs in which nuclear accumulation of beta-catenin were observed, which may suggest that MARKL1 plays some role in hepatocellular carcinogenesis.