Merck
  • Isolation of TAO1, a protein kinase that activates MEKs in stress-activated protein kinase cascades.
Isolation of TAO1, a protein kinase that activates MEKs in stress-activated protein kinase cascades.
The Journal of biological chemistry (1998-10-24)
M Hutchison, K S Berman, M H Cobb
ZUSAMMENFASSUNG
Several components of the budding yeast pheromone-response pathway are conserved in mammalian mitogen-activated protein (MAP) kinase pathways. Thus, we used degenerate oligonucleotides derived from the sequence of the Saccharomyces cerevisiae protein kinase Ste20p to amplify related sequences from the rat. One of these sequences was used to clone a rat Ste20p homolog, which we called TAO1 for its one thousand and one amino acids. Northern analysis shows TAO1 is highly expressed in brain, as is a homolog TAO2. Recombinant TAO1 was expressed and purified from Sf9 cells. In vitro, it activated MAP/extracellular signal-regulated protein kinase (ERK) kinases (MEKs) 3, 4, and 6 of the stress-responsive MAP kinase pathways, but not MEK1 or 2 of the classical MAP kinase pathway. TAO1 activated MEK3 but not MEK4 or MEK6 in transfected cells. MEK3 coimmunoprecipitated with TAO1 when they were expressed in 293 cells. In addition, immunoreactive MEK3 endogenous to Sf9 cells copurified with TAO1 produced from a recombinant baculovirus. The activation of and binding to MEK3 by TAO1 implicates TAO1 in the regulation of the p38-containing stress-responsive MAP kinase pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
TAOK1 (1-314), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
TAO1 Protein, active, 10 µg, Active, N-terminal 6His-tagged, recombinant, human TAO1 amino acids 1-327. For use in Kinase Assays.
Sigma-Aldrich
TAOK2 (1-314), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution