Merck
AIM2 inflammasome is activated by pharmacological disruption of nuclear envelope integrity.
Proceedings of the National Academy of Sciences of the United States of America (2016-07-28)
Antonia Di Micco, Gianluca Frera, Jérôme Lugrin, Yvan Jamilloux, Erh-Ting Hsu, Aubry Tardivel, Aude De Gassart, Léa Zaffalon, Bojan Bujisic, Stefanie Siegert, Manfredo Quadroni, Petr Broz, Thomas Henry, Christine A Hrycyna, Fabio Martinon
ZUSAMMENFASSUNG
Inflammasomes are critical sensors that convey cellular stress and pathogen presence to the immune system by activating inflammatory caspases and cytokines such as IL-1β. The nature of endogenous stress signals that activate inflammasomes remains unclear. Here we show that an inhibitor of the HIV aspartyl protease, Nelfinavir, triggers inflammasome formation and elicits an IL-1R-dependent inflammation in mice. We found that Nelfinavir impaired the maturation of lamin A, a structural component of the nuclear envelope, thereby promoting the release of DNA in the cytosol. Moreover, deficiency of the cytosolic DNA-sensor AIM2 impaired Nelfinavir-mediated inflammasome activation. These findings identify a pharmacologic activator of inflammasome and demonstrate the role of AIM2 in detecting endogenous DNA release upon perturbation of nuclear envelope integrity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bernsteinsäure bis(N-Hydroxysuccinimid-Ester), ≥95%, powder