Merck
  • "Reduction of carbonyl compounds promoted by silicon hydrides under the influence of trimethylsilyl-based reagent"
"Reduction of carbonyl compounds promoted by silicon hydrides under the influence of trimethylsilyl-based reagent"
Canadian Journal of Chemistry (1986)
Aizpurua.MJ, et al.