Merck
  • The GCN5-CITED2-PKA signalling module controls hepatic glucose metabolism through a cAMP-induced substrate switch.
The GCN5-CITED2-PKA signalling module controls hepatic glucose metabolism through a cAMP-induced substrate switch.
Nature communications (2016-11-23)
Mashito Sakai, Tomoko Tujimura-Hayakawa, Takashi Yagi, Hiroyuki Yano, Masaru Mitsushima, Hiroyuki Unoki-Kubota, Yasushi Kaburagi, Hiroshi Inoue, Yoshiaki Kido, Masato Kasuga, Michihiro Matsumoto
ZUSAMMENFASSUNG
Hepatic gluconeogenesis during fasting results from gluconeogenic gene activation via the glucagon-cAMP-protein kinase A (PKA) pathway, a process whose dysregulation underlies fasting hyperglycemia in diabetes. Such transcriptional activation requires epigenetic changes at promoters by mechanisms that have remained unclear. Here we show that GCN5 functions both as a histone acetyltransferase (HAT) to activate fasting gluconeogenesis and as an acetyltransferase for the transcriptional co-activator PGC-1α to inhibit gluconeogenesis in the fed state. During fasting, PKA phosphorylates GCN5 in a manner dependent on the transcriptional coregulator CITED2, thereby increasing its acetyltransferase activity for histone and attenuating that for PGC-1α. This substrate switch concomitantly promotes both epigenetic changes associated with transcriptional activation and PGC-1α-mediated coactivation, thereby triggering gluconeogenesis. The GCN5-CITED2-PKA signalling module and associated GCN5 substrate switch thus serve as a key driver of gluconeogenesis. Disruption of this module ameliorates hyperglycemia in obese diabetic animals, offering a potential therapeutic strategy for such conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
8-(4-Chlorphenylthio)adenosin-3′,5′-zyklisches Monophosphat Natriumsalz, ≥97.0% (HPLC), powder
Sigma-Aldrich
Anti-Trimethyl-Histon H3(Lys4)-Antikörper, Klon 15-10C-E4, Kaninchen, monoklonal, clone 15-10C-E4, Upstate®, from rabbit
Sigma-Aldrich
Anti-p300 CT-Antikörper, Klon RW128, clone RW128, Upstate®, from mouse