Merck
  • Molecular Mechanism of Action of Antimalarial Benzoisothiazolones: Species-Selective Inhibitors of the Plasmodium spp. MEP Pathway enzyme, IspD.
Molecular Mechanism of Action of Antimalarial Benzoisothiazolones: Species-Selective Inhibitors of the Plasmodium spp. MEP Pathway enzyme, IspD.
Scientific reports (2016-11-20)
Kathryn E Price, Christopher M Armstrong, Leah S Imlay, Dana M Hodge, C Pidathala, Natalie J Roberts, Jooyoung Park, Marwa Mikati, Raman Sharma, Alexandre S Lawrenson, Niraj H Tolia, Neil G Berry, Paul M O'Neill, Audrey R Odom John
ZUSAMMENFASSUNG
The methylerythritol phosphate (MEP) pathway is an essential metabolic pathway found in malaria parasites, but absent in mammals, making it a highly attractive target for the discovery of novel and selective antimalarial therapies. Using high-throughput screening, we have identified 2-phenyl benzo[d]isothiazol-3(2H)-ones as species-selective inhibitors of Plasmodium spp. 2-C-methyl-
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Isopentenylpyrophosphat Triammoniumsalz -Lösung, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)