Merck
Anticodon-like binding of the HIV-1 tRNA-like element to human lysyl-tRNA synthetase.
RNA (New York, N.Y.) (2016-11-18)
Sheng Liu, Roopa Comandur, Christopher P Jones, Pearl Tsang, Karin Musier-Forsyth
ZUSAMMENFASSUNG
A critical step in the HIV-1 lifecycle involves reverse transcription of the viral genomic RNA (gRNA). Human tRNA
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluoreszein-5-thiosemicarbazid, suitable for fluorescence, ~80% (HPCE)