Merck
  • Anti-neuroinflammatory effect of Sophoraflavanone G from Sophora alopecuroides in LPS-activated BV2 microglia by MAPK, JAK/STAT and Nrf2/HO-1 signaling pathways.
Anti-neuroinflammatory effect of Sophoraflavanone G from Sophora alopecuroides in LPS-activated BV2 microglia by MAPK, JAK/STAT and Nrf2/HO-1 signaling pathways.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2016-11-09)
Chao Guo, Lei Yang, Chuan-Xing Wan, Yuan-Zheng Xia, Chao Zhang, Meng-Han Chen, Zhen-Dong Wang, Zhong-Rui Li, Xue-Mei Li, Ya-Di Geng, Ling-Yi Kong
ZUSAMMENFASSUNG
Neuroinflammation plays a vital role in Alzheimer's disease (AD) and other neurodegenerative conditions. Sophora alopecuroides is widely used in traditional Uighur's medicine for the treatment of inflammation. Sophoraflavanone G (SG), a major flavonoid found in the S. alopecuroides, has also been reported to exhibit anti-inflammatory activity both in vitro and in vivo. However, the effect of S. alopecuroides and SG on microglia-mediated neuroinflammation has not been investigated. The present study was designed to evaluate the anti-neuroinflammatory effect of S. alopecuroides and SG against lipopolysaccharide (LPS)-activated BV2 microglial cells and to explore the underlying mechanisms. We measured the production of pro-inflammatory mediators and cytokines, and analyzed relevant mRNA and protein expressions by qRT-PCR and Western Blot. S. alopecuroides extract (SAE) and SG inhibited the LPS-induced release of nitric oxide (NO), prostaglandin E S. alopecuroides and SG displayed anti-neuroinflammatory activity in LPS-activated BV2 microglia. SG was able to inhibit the neuroinflammation by MAPKs, PI3K/AKT, JAK/STAT and Nrf2/HO-1 signaling pathways and might act as a natural therapeutic agent to be further developed for the treatment of various neuroinflammatory conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SP600125, ≥98% (HPLC)