Merck
Wnt5a induces renal AQP2 expression by activating calcineurin signalling pathway.
Nature communications (2016-11-29)
Fumiaki Ando, Eisei Sohara, Tetsuji Morimoto, Naofumi Yui, Naohiro Nomura, Eriko Kikuchi, Daiei Takahashi, Takayasu Mori, Alain Vandewalle, Tatemitsu Rai, Sei Sasaki, Yoshiaki Kondo, Shinichi Uchida
ZUSAMMENFASSUNG
Heritable nephrogenic diabetes insipidus (NDI) is characterized by defective urine concentration mechanisms in the kidney, which are mainly caused by loss-of-function mutations in the vasopressin type 2 receptor. For the treatment of heritable NDI, novel strategies that bypass the defective vasopressin type 2 receptor are required to activate the aquaporin-2 (AQP2) water channel. Here we show that Wnt5a regulates AQP2 protein expression, phosphorylation and trafficking, suggesting that Wnt5a is an endogenous ligand that can regulate AQP2 without the activation of the classic vasopressin/cAMP signalling pathway. Wnt5a successfully increases the apical membrane localization of AQP2 and urine osmolality in an NDI mouse model. We also demonstrate that calcineurin is a key regulator of Wnt5a-induced AQP2 activation without affecting intracellular cAMP level and PKA activity. The importance of calcineurin is further confirmed with its activator, arachidonic acid, which shows vasopressin-like effects underlining that calcineurin activators may be potential therapeutic targets for heritable NDI.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Y-27632 -dihydrochlorid, ≥98% (HPLC)
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
H-89 dihydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Arachidonsäure-Natriumsalz aus Mortierella alpina, ≥98.5% (GC)
Sigma-Aldrich
KN-93, ≥98% (HPLC)
Sigma-Aldrich
N-(6-Aminohexyl)-5-chlor-1-Naphthalinsulfonamid -hydrochlorid
Sigma-Aldrich
Anti-Frizzled 5 Antibody, Upstate®, from rabbit