- Acyclic nucleoside phosphonates containing the amide bond.
Acyclic nucleoside phosphonates containing the amide bond.
Monatshefte fur chemie (2016-11-25)
Iwona E Głowacka, Dorota G Piotrowska, Graciela Andrei, Dominique Schols, Robert Snoeck, Andrzej E Wróblewski
PMID27881885
ZUSAMMENFASSUNG
To study the influence of a linker rigidity and donor-acceptor properties, the P-CH
MATERIALIEN