Merck
  • Efficient replacement of plasma membrane outer leaflet phospholipids and sphingolipids in cells with exogenous lipids.
Efficient replacement of plasma membrane outer leaflet phospholipids and sphingolipids in cells with exogenous lipids.
Proceedings of the National Academy of Sciences of the United States of America (2016-11-23)
Guangtao Li, JiHyun Kim, Zhen Huang, Johnna R St Clair, Deborah A Brown, Erwin London
ZUSAMMENFASSUNG
Our understanding of membranes and membrane lipid function has lagged far behind that of nucleic acids and proteins, largely because it is difficult to manipulate cellular membrane lipid composition. To help solve this problem, we show that methyl-α-cyclodextrin (MαCD)-catalyzed lipid exchange can be used to maximally replace the sphingolipids and phospholipids in the outer leaflet of the plasma membrane of living mammalian cells with exogenous lipids, including unnatural lipids. In addition, lipid exchange experiments revealed that 70-80% of cell sphingomyelin resided in the plasma membrane outer leaflet; the asymmetry of metabolically active cells was similar to that previously defined for erythrocytes, as judged by outer leaflet lipid composition; and plasma membrane outer leaflet phosphatidylcholine had a significantly lower level of unsaturation than phosphatidylcholine in the remainder of the cell. The data also provided a rough estimate for the total cellular lipids residing in the plasma membrane (about half). In addition to such lipidomics applications, the exchange method should have wide potential for investigations of lipid function and modification of cellular behavior by modification of lipids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Sigma-Aldrich
1,2-Dioleoyl-sn-Glycero-3-Phosphocholin, lyophilized powder
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
Cholesterol (ovine), Avanti Research - A Croda Brand
Avanti
16:0–18:1 PE, Avanti Research - A Croda Brand
Avanti
Brain SM, Avanti Research - A Croda Brand
Avanti
16:0–18:1 PS (POPS), Avanti Research - A Croda Brand
Avanti
16:0–18:1 PS (POPS), Avanti Research - A Croda Brand
Avanti
Egg SM, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PE, Avanti Research - A Croda Brand
Avanti
Eier-SM, Avanti Research - A Croda Brand
Avanti
Brain SM, Avanti Research - A Croda Brand
Avanti
16:0 NBD PE, Avanti Research - A Croda Brand, powder
Avanti
Milk SM, Avanti Research - A Croda Brand
Avanti
16:0 NBD PE, Avanti Research - A Croda Brand 810144C
Avanti
Milk SM, Avanti Research - A Croda Brand
Avanti
17:0 SM (d18:1/17:0), Avanti Research - A Croda Brand 860585P, powder