Merck
Evaluation of genetic diversity of
Physiology and molecular biology of plants : an international journal of functional plant biology (2016-12-08)
Noor Zafirah Ismail, Hasni Arsad, Mohammed Razip Samian, Abdul Hafiz Ab Majid, Mohammad Razak Hamdan
ZUSAMMENFASSUNG
Three polymerase chain reaction (PCR) techniques were compared to analyse the genetic diversity of
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-(1-Pyridin)-1-propansulfonat, ≥97.0% (N)