Merck
  • Polycyclic aromatic hydrocarbons are associated with increased risk of chronic obstructive pulmonary disease during haze events in China.
Polycyclic aromatic hydrocarbons are associated with increased risk of chronic obstructive pulmonary disease during haze events in China.
The Science of the total environment (2016-09-12)
Lingyan Yang, Wen-Cheng Wang, Shih-Chun Candice Lung, Zhelin Sun, Chongjun Chen, Jen-Kun Chen, Qiang Zou, Yu-Hsin Lin, Chia-Hua Lin
ZUSAMMENFASSUNG
Although exposure to particulate matter with a diameter of <2.5μm (PM
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2′,7′-Dichlorfluorescein-Diacetat, ≥97%