Merck
  • Non-imidazole-based histamine H3 receptor antagonists with anticonvulsant activity in different seizure models in male adult rats.
Non-imidazole-based histamine H3 receptor antagonists with anticonvulsant activity in different seizure models in male adult rats.
Drug design, development and therapy (2016-12-10)
Bassem Sadek, Ali Saad, Gniewomir Latacz, Kamil Kuder, Agnieszka Olejarz, Tadeusz Karcz, Holger Stark, Katarzyna Kieć-Kononowicz
ZUSAMMENFASSUNG
A series of twelve novel non-imidazole-based ligands (
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Pyroglutaminsäure-2-naphthylamid, ≥99% (TLC)
Sigma-Aldrich
Pyrrolidon -hydrotribromid, 97%