Merck
  • Protective effects of the roots of Angelica sinensis on strenuous exercise-induced sports anemia in rats.
Protective effects of the roots of Angelica sinensis on strenuous exercise-induced sports anemia in rats.
Journal of ethnopharmacology (2016-08-09)
Chih-Wei Chang, Yi-Ming Chen, Yi-Ju Hsu, Chi-Chang Huang, Yu-Tse Wu, Mei-Chich Hsu
ZUSAMMENFASSUNG
Sports anemia is a persistent and severe problem in athletes owing to strenuous exercise-induced oxidative stress and hepcidin upregulation. The roots of Angelica sinensis (AS), a familiar traditional Chinese medicine, has been used for replenishing blood since antiquity. To evaluate the effects of ethanolic AS extract in a 4-week study on sports anemia in female Wistar rats. To induce anemia, a strenuous exercise protocol consisting of running and swimming was employed with increasing intensity. Animals were randomly assigned to the following groups: control group; strenuous exercise group; and strenuous exercise and AS extract-treated group (300mgkg AS extract (consisting of ferulic acid, Z-ligustilide, and n-butylidenephthalide) treatment improved forelimb grip strength and rescued exercise-induced anemia by significantly elevating the red blood cell counts and hemoglobin concentrations as well as hematocrit levels (p<0.05). AS modulated the iron metabolism through decreasing serum hepcidin-25 concentrations by 33.0% (p<0.05) and increasing serum iron levels by 34.3% (p<0.01). The hepatic injury marker serum alanine aminotransferase concentrations were also reduced, followed by increased antioxidant enzyme catalase expression in the liver (p<0.05). Furthermore, substantial attenuation of hepatic interleukin-6 expression and preservation of muscle glycogen content suggested the additional roles of AS acting on sports anemia and physical performance. Our findings evidenced a novel and promising therapeutic approach for AS treatment for rescuing the anemic condition induced following 4 weeks of strenuous exercise.