Merck
Quantum chemical study of a derivative of 3-substituted dithiocarbamic flavanone.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2016-04-28)
Steluta Gosav, Nicoleta Paduraru, Dan Maftei, Mihail Lucian Birsa, Mirela Praisler
ZUSAMMENFASSUNG
The aim of this work is to characterize a quite novel 3-dithiocarbamic flavonoid by vibrational spectroscopy in conjunction with Density Functional Theory (DFT) calculations. Quantum mechanics calculations of energies, geometries and vibrational wavenumbers in the ground state were carried out by using hybrid functional B3LYP with 6-311G(d,p) as basis set. The results indicate a remarkable agreement between the calculated molecular geometries, as well as vibrational frequencies, and the corresponding experimental data. In addition, a complete assignment of all the absorption bands present in the vibrational spectrum has been performed. In order to assess its chemical potential, quantum molecular descriptors characterizing the interactions between the 3-dithiocarbamic flavonoid and its biological receptors have been computed. The frontier molecular orbitals and the HOMO-LUMO energy gap have been used in order to explain the way in which the new molecule can interact with other species and to characterize its molecular chemical stability/reactivity. The molecular electrostatic potential (MEP) map, computed in order to identify the sites of the studied flavonoid that are most likely to interact with electrophilic and nucleophilic species, is discussed.