Merck
  • DAAM1 stabilizes epithelial junctions by restraining WAVE complex-dependent lateral membrane motility.
DAAM1 stabilizes epithelial junctions by restraining WAVE complex-dependent lateral membrane motility.
The Journal of cell biology (2016-11-04)
Tamako Nishimura, Shoko Ito, Hiroko Saito, Sylvain Hiver, Kenta Shigetomi, Junichi Ikenouchi, Masatoshi Takeichi
ZUSAMMENFASSUNG
Epithelial junctions comprise two subdomains, the apical junctional complex (AJC) and the adjacent lateral membrane contacts (LCs), that span the majority of the junction. The AJC is lined with circumferential actin cables, whereas the LCs are associated with less-organized actin filaments whose roles are elusive. We found that DAAM1, a formin family actin regulator, accumulated at the LCs, and its depletion caused dispersion of actin filaments at these sites while hardly affecting circumferential actin cables. DAAM1 loss enhanced the motility of LC-forming membranes, leading to their invasion of neighboring cell layers, as well as disruption of polarized epithelial layers. We found that components of the WAVE complex and its downstream targets were required for the elevation of LC motility caused by DAAM1 loss. These findings suggest that the LC membranes are motile by nature because of the WAVE complex, but DAAM1-mediated actin regulation normally restrains this motility, thereby stabilizing epithelial architecture, and that DAAM1 loss evokes invasive abilities of epithelial cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human WASF2
Sigma-Aldrich
MISSION® esiRNA, targeting human DAAM1