Merck
  • Significant Phylogenetic Signal and Climate-Related Trends in Leaf Caloric Value from Tropical to Cold-Temperate Forests.
Significant Phylogenetic Signal and Climate-Related Trends in Leaf Caloric Value from Tropical to Cold-Temperate Forests.
Scientific reports (2016-11-20)
Guangyan Song, Ying Li, Jiahui Zhang, Meiling Li, Jihua Hou, Nianpeng He
ZUSAMMENFASSUNG
Leaf caloric value (LCV) is a useful index to represent the conversion efficiency of leaves for solar energy. We investigated the spatial pattern of LCV and explored the factors (phylogeny, climate, and soil) that influence them at a large scale by determining LCV standardized by leaf area in 920 plant species from nine forest communities along the 3700 km North-South Transect of Eastern China. LCV ranged from 0.024 to 1.056 kJ cm