- Enhanced Ascorbate Regeneration Via Dehydroascorbate Reductase Confers Tolerance to Photo-Oxidative Stress in Chlamydomonas reinhardtii.
Enhanced Ascorbate Regeneration Via Dehydroascorbate Reductase Confers Tolerance to Photo-Oxidative Stress in Chlamydomonas reinhardtii.
Plant & cell physiology (2016-07-22)
Shu-Tseng Lin, Chih-Wen Chiou, Yen-Lin Chu, Yu Hsiao, Yu-Fei Tseng, Yi-Chun Chen, Hsien-Jung Chen, Hsin-Yang Chang, Tse-Min Lee
PMID27440549
ZUSAMMENFASSUNG
The role of ascorbate (AsA) recycling via dehydroascorbate reductase (DHAR) in the tolerance of Chlamydomonas reinhardtii to photo-oxidative stress was examined. The activity of DHAR and the abundance of the CrDHAR1 (Cre10.g456750) transcript increased after moderate light (ML; 750 µmol m