Merck
  • Implementation of output prediction models for a passively double-scattered proton therapy system.
Implementation of output prediction models for a passively double-scattered proton therapy system.
Medical physics (2016-11-04)
Sven Ferguson, Salahuddin Ahmad, Hosang Jin
ZUSAMMENFASSUNG
Two output (cGy/MU) prediction models (one existing and one newly developed) for a passively double-scattered proton therapy system are implemented and investigated for clinical use. Variations of each model are tested for accuracy in order to determine the most viable prediction model. The first output prediction model [model (1)] is a semianalytical model proposed by Kooy et al. [Phys. Med. Biol. 50, 5847-5856 (2005)], which employs three main factors. The first factor (basic output prediction) uses a unique combined parameter [r = (R - M)/M] of range (R) and modulation [M; spread-out Bragg peak (SOBP) width] along with option specific fitting parameters. The second factor takes into account minor source shifts using a linear fit due to varying beamline configurations for different options. The final factor accounts for a condition where the point of measurement is not at the isocenter or away from the middle of the SOBP based on an inverse-square correction. The second model [model (2)] is a novel quartic polynomial fit of the basic output prediction whose idea was inspired by the first model. Different variations in the definition of R and M at distal (D) and proximal (P) ends resulted in the exploration of three variations of r for both models: r The mean differences between measurements and predictions ([predicted - measured]/measured × 100%) were -0.41% ± 1.78% (r The first existing model has proven to be a successful predictor of output for our compact double-scattering proton therapy system. The new model performed comparably to the first model and showed better performance in some options due to a great degree of flexibility of a polynomial fit. Both models performed well using r
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent