Merck
Sustained Gq-Protein Signaling Disrupts Striatal Circuits via JNK.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2016-10-14)
Luigi Bellocchio, Andrea Ruiz-Calvo, Anna Chiarlone, Magali Cabanas, Eva Resel, Jean-René Cazalets, Cristina Blázquez, Yoon H Cho, Ismael Galve-Roperh, Manuel Guzmán
ZUSAMMENFASSUNG
The dorsal striatum is a major input structure of the basal ganglia and plays a key role in the control of vital processes such as motor behavior, cognition, and motivation. The functionality of striatal neurons is tightly controlled by various metabotropic receptors. Whereas the Gs/Gi-protein-dependent tuning of striatal neurons is fairly well known, the precise impact and underlying mechanism of Gq-protein-dependent signals remain poorly understood. Here, using different experimental approaches, especially designer receptor exclusively activated by designer drug (DREADD) chemogenetic technology, we found that sustained activation of Gq-protein signaling impairs the functionality of striatal neurons and we unveil the precise molecular mechanism underlying this process: a phospholipase C/Ca2+/proline-rich tyrosine kinase 2/cJun N-terminal kinase pathway. Moreover, engagement of this intracellular signaling route was functionally active in the mouse dorsal striatum in vivo, as proven by the disruption of neuronal integrity and behavioral tasks. To analyze this effect anatomically, we manipulated Gq-protein-dependent signaling selectively in neurons belonging to the direct or indirect striatal pathway. Acute Gq-protein activation in direct-pathway or indirect-pathway neurons produced an enhancement or a decrease, respectively, of activity-dependent parameters. In contrast, sustained Gq-protein activation impaired the functionality of direct-pathway and indirect-pathway neurons and disrupted the behavioral performance and electroencephalography-related activity tasks controlled by either anatomical framework. Collectively, these findings define the molecular mechanism and functional relevance of Gq-protein-driven signals in striatal circuits under normal and overactivated states. The dorsal striatum is a major input structure of the basal ganglia and plays a key role in the control of vital processes such as motor behavior, cognition, and motivation. Whereas the Gs/Gi-protein-dependent tuning of striatal neurons is fairly well known, the precise impact and underlying mechanism of Gq-protein-dependent signals remain unclear. Here, we show that striatal circuits can be "turned on" by acute Gq-protein signaling or "turned off" by sustained Gq-protein signaling. Specifically, sustained Gq-protein signaling inactivates striatal neurons by an intracellular pathway that relies on cJun N-terminal kinase. Overall, this study sheds new light onto the molecular mechanism and functional relevance of Gq-protein-driven signals in striatal circuits under normal and overactivated states.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Cholin-Acetyltransferase-Antikörper, Chemicon®, from goat
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Monoklonaler Anti-β-Tubulin-III-Antikörper in Maus hergestellte Antikörper, clone SDL.3D10, ascites fluid
Sigma-Aldrich
SP600125, ≥98% (HPLC)