Merck
  • Use of focused ultrasonication in activity-based profiling of deubiquitinating enzymes in tissue.
Use of focused ultrasonication in activity-based profiling of deubiquitinating enzymes in tissue.
Analytical biochemistry (2016-10-22)
Bindu Nanduri, Leslie A Shack, Aswathy N Rai, William B Epperson, Wes Baumgartner, Ty B Schmidt, Mariola J Edelmann
ZUSAMMENFASSUNG
To develop a reproducible tissue lysis method that retains enzyme function for activity-based protein profiling, we compared four different methods to obtain protein extracts from bovine lung tissue: focused ultrasonication, standard sonication, mortar & pestle method, and homogenization combined with standard sonication. Focused ultrasonication and mortar & pestle methods were sufficiently effective for activity-based profiling of deubiquitinases in tissue, and focused ultrasonication also had the fastest processing time. We used focused-ultrasonicator for subsequent activity-based proteomic analysis of deubiquitinases to test the compatibility of this method in sample preparation for activity-based chemical proteomics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Influenza-Hämagglutinin(HA)-Peptid, ≥97% (HPLC)