Merck
  • Molecular docking studies and synthesis of novel bisbenzimidazole derivatives as inhibitors of α-glucosidase.
Molecular docking studies and synthesis of novel bisbenzimidazole derivatives as inhibitors of α-glucosidase.
Bioorganic & medicinal chemistry (2016-09-01)
Musa Özil, Mustafa Emirik, Ali Beldüz, Serdar Ülker
ZUSAMMENFASSUNG
A series of bisbenzimidazole derivatives starting from o-phenylenediamine and 4-nitro-o-phenylenediamine were prepared with oxalic acid. Most of the reactions were conducted using both the microwave and conventional methods to compare yields and reaction times. The operational simplicity, environmental friendly conditions and high yield in a significantly short reaction time were the major benefits. All substances' inhibitory activities against α-glucosidase were evaluated. The results may suggest a significant role for the nature of bisbenzimidazole compounds in their inhibitory action against α-glucosidase. They showed different range of α-glucosidase inhibitory potential with IC
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Propylene sulfite, 99%